Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-13 (Collagenase-3) Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS825P1ABX

 View more versions of this product

Catalog No. MS825P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Antigen MMP-13 (Collagenase-3) Ab-1
Regulatory Status RUO
Format Concentrated
Clone VIIIA2
Immunogen Human recombinant collagenase-3 protein
Isotype IgG2a kappa
Quantity 100µL
Applications Affinity Purification
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Host Species Mouse
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Proteases And Inhibitors
Concentration 1mg/mL
Type Primary
Classification Monoclonal

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-1, Mouse Monoclonal Antibody.

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Mouse

Clone: VIIIA2

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Human recombinant collagenase-3 protein

Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human chondrosarcoma SW1353 cells; Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation (Native verified)
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO