Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-10 (Stromelysin-2) Ab-2, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ MMP-10 (Stromelysin-2) Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS822P1ABX

 View more versions of this product

Catalog No. MS822P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Format Concentrated
Immunogen Human recombinant MMP-10 (stromelysin-2 protein)
Quantity 100µL
Regulatory Status RUO
Clone IVC5
Antigen MMP-10 (Stromelysin-2) Ab-2
Host Species Mouse
Isotype IgG2b
Research Discipline Proteases And Inhibitors
Type Primary
Concentration 1mg/mL
Classification Monoclonal
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ MMP-10 (Stromelysin-2) Ab-2, Mouse Monoclonal Antibody.

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading extracellular matrix. Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The two forms of stromelysin (SL), types 1 (MMP-3) and 2 (MMP-10), share 82% sequence homology, but exhibit differences in cellular synthesis and inducibility by cytokines and growth factors in vitro.

Host Species: Mouse

Clone: IVC5

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human recombinant MMP-10 (stromelysin-2 protein)

Molecular Weight: ∽57kDa (proform) and ∽44kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human large cell lymphoma U937 cells; Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO