missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-1 (Collagenase-1) Ab-6, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ MMP-1 (Collagenase-1) Ab-6, Rabbit Polyclonal Antibody.

$295.73 - $1,414.02

Specifications

Antigen MMP-1 (Collagenase-1) Ab-6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB1536P Thermo Scientific™ Lab Vision™
RB1536P
Purified Ab with BSA and Azide 1mL Each for $1,287.33
RB1536P0 Thermo Scientific™ Lab Vision™
RB1536P0
Purified Ab with BSA and Azide 100μL Each for $295.73
RB1536P1 Thermo Scientific™ Lab Vision™
RB1536P1
Purified Ab with BSA and Azide 500μL Each for $779.04
RB1536P1ABX Thermo Scientific™ Lab Vision™
RB1536P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $865.12
RB1536PABX Thermo Scientific™ Lab Vision™
RB1536PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,414.02
RB1536R7 Thermo Scientific™ Lab Vision™
RB1536R7
Ready-to-use for IHC (UVLP) 7mL Each for $673.04
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ MMP-1 (Collagenase-1) Ab-6, Rabbit Polyclonal Antibody.

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. Interstitial collagenase (MMP-1), 72kDa gelatinase A/type IV collagenase (MMP-2), and 92kDa gelatinase B/type IV collagenase (MMP-9) dissolve extracellular matrix (ECM) and may initiate and promote angiogenesis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Does not react with cow. Others not known.

Epitope: Middle region of MMP-1

Immunogen: A synthetic peptide from the middle region of the human MMP-1

Molecular Weight: 52kDa (unglycosylated) and 57kDa (glycosylated) species of proform of MMP-1 plus 42kDa (unglycosylated) and 47kDa (glycosylated) active form of MMP-1

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells; Placenta; Bladder, breast, ovarian, colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

MMP-1 (Collagenase-1) Ab-6
Polyclonal
Human
RUO
Primary
IgG
Immunohistochemistry (Paraffin)
Unlabeled
Rabbit
Proteases And Inhibitors
A synthetic peptide from the middle region of the human MMP-1