Please login to your online account to display your discounted pricing

MM 77 dihydrochloride, Tocris


5-HT1A (postsynaptic) antagonist

Quantity 10mg

Manufacturer: r & d systems  093310

 View more versions of this product

Catalog No. 093310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 418.36
Chemical Name or Material MM 77 dihydrochloride
Molecular Formula C19H27N3O3·2HCl
CAS 159187-70-9
Synonym 1-(2-Methoxyphenyl)-4-(4-succinimidobutyl)piperazine dihydrochloride
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material MM 77 dihydrochloride
Assay Percent Range >98%
Molecular Formula C19H27N3O3·2HCl
Formula Weight 418.36
Synonym 1-(2-Methoxyphenyl)-4-(4-succinimidobutyl)piperazine dihydrochloride
Accessories

Store at Room Temperature