Please login to your online account to display your discounted pricing

MM 77 dihydrochloride, Tocris

5-HT1A (postsynaptic) antagonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material MM 77 dihydrochloride
Assay Percent Range >98%
Molecular Formula C19H27N3O3·2HCl
Formula Weight 418.36
Synonym 1-(2-Methoxyphenyl)-4-(4-succinimidobutyl)piperazine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

093350

 
r & d systems
093350
50mg Each for $762.58

093310

 
r & d systems
093310
10mg Each for $181.19
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 418.36
Chemical Name or Material MM 77 dihydrochloride
Assay Percent Range >98%
CAS 159187-70-9
Molecular Formula C19H27N3O3·2HCl
Synonym 1-(2-Methoxyphenyl)-4-(4-succinimidobutyl)piperazine dihydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature