Please login to your online account to display your discounted pricing

MLR 1023, Tocris™

Selective allosteric activator of Lyn kinase

$212.74 - $886.68

Chemical Identifiers

Chemical Name or Material MLR 1023
Assay Percent Range >99%
Molecular Formula C11H10N2O2
Formula Weight 202.21
Synonym 5-(3-Methylphenoxy)-2(1H)-pyrimidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

458250

 
r & d systems
458250
50mg Each for $886.68

458210

 
r & d systems
458210
10mg Each for $212.74
Description & Specifications

Specifications

Formula Weight 202.21
Quantity 50mg
Chemical Name or Material MLR 1023
Assay Percent Range >99%
Molecular Formula C11H10N2O2
CAS 41964-07-2
Synonym 5-(3-Methylphenoxy)-2(1H)-pyrimidinone
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C