Invitrogen™ MLCK (MLCK2) Recombinant Human Protein DFS Item
MLCK (MLCK2) is now also known as MYLK3.
Manufacturer:  Invitrogen™ LSPV4023
 View more versions of this product
Catalog No. PV4023
Description
Description
MLCK (MLCK2) is of of the myosin regulatory light chain kinases which facilitates muscle contraction.
Specifications
Specifications
NP_872299, R175G
Frozen
113.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
MLCK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MLCK (MLCK2)
Baculovirus
Human
GST Tag
Documents