Please login to your online account to display your discounted pricing

ML 9 hydrochloride, Tocris

Myosin light chain kinase inhibitor

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material ML 9 hydrochloride
Assay Percent Range >99%
Molecular Formula C15H17ClN2O2S·HCl
Formula Weight 361.29
Synonym 1-(5-Chloronaphthalenesulfonyl)homopiperazine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

043110

 
r & d systems
043110
10mg Each for $181.19

043150

 
r & d systems
043150
50mg Each for $762.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 361.29
Chemical Name or Material ML 9 hydrochloride
CAS 105637-50-1
Synonym 1-(5-Chloronaphthalenesulfonyl)homopiperazine hydrochloride
Assay Percent Range >99%
Molecular Formula C15H17ClN2O2S·HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature