Please login to your online account to display your discounted pricing

ML 3403, Tocris™

p38 inhibitor

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material ML 3403
Assay Percent Range >98%
Molecular Formula C23H21FN4S
Formula Weight 404.5
Synonym [5-[4-(4-Fluorophenyl)-2-methylthio-1H-imidazol-4-yl]-N-(1-phenylethyl)-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

458650

 
r & d systems
RDS458650
50mg Each for $1,073.10

458610

 
r & d systems
RDS458610
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 404.5
Quantity 50mg
Chemical Name or Material ML 3403
Assay Percent Range >98%
Molecular Formula C23H21FN4S
CAS 549505-65-9
Synonym [5-[4-(4-Fluorophenyl)-2-methylthio-1H-imidazol-4-yl]-N-(1-phenylethyl)-2-pyridinamine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C