missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ML 298 hydrochloride, Tocris™

Selective PLD2 inhibitor

$218.74 - $915.10

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C22H23F3N4O2·HCl
Formula Weight 468.9
Synonym 3,4-Difluoro-N-[2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]dec-8-yl]ethyl]benzamide hydrochloride
Chemical Name or Material ML 298 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
489510 R & D Systems
RDS489510
10mg Each for $218.74
489550 R & D Systems
RDS489550
50mg Each for $915.10
Specifications Description & Specifications

Chemical Identifiers

>98%
468.9
ML 298 hydrochloride
C22H23F3N4O2·HCl
3,4-Difluoro-N-[2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]dec-8-yl]ethyl]benzamide hydrochloride

Specifications

10mg
C22H23F3N4O2·HCl
3,4-Difluoro-N-[2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]dec-8-yl]ethyl]benzamide hydrochloride
>98%
468.9
ML 298 hydrochloride

Safety and Handling

Recommended Storage : Desiccate at Room Temperature