Please login to your online account to display your discounted pricing

ML 218 hydrochloride, Tocris™

Ca2+ channel blocker (T-type)

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material ML 218 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H26Cl2N2O·HCl
Formula Weight 405.79
Synonym 3,5-Dichloro-N-[[(1α,5α,6-exo,6α)-3-(3,3-dimethylbutyl)-3-azabicyclo[3.1.0]hex-6-yl]methyl]-benzamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

450750

 
r & d systems
RDS450750
50mg Each for $1,050.42

450710

 
r & d systems
RDS450710
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 405.79
Quantity 50mg
Chemical Name or Material ML 218 hydrochloride
Molecular Formula C19H26Cl2N2O·HCl
Synonym 3,5-Dichloro-N-[[(1α,5α,6-exo,6α)-3-(3,3-dimethylbutyl)-3-azabicyclo[3.1.0]hex-6-yl]methyl]-benzamide hydrochloride
Recommended Storage Store at +4°C
Assay Percent Range >99%
CAS 1346233-68-8
Safety and Handling

Store at +4°C