Please login to your online account to display your discounted pricing

ML 204, Tocris™

Selective TRPC4 blocker

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material ML 204
Assay Percent Range >99%
Molecular Formula C15H18N2
Formula Weight 226.32
Synonym 4-Methyl-2-(1-piperidinyl)quinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

473210

 
r & d systems
RDS473210
10mg Each for $177.37

473250

 
r & d systems
RDS473250
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 226.32
Quantity 10mg
Assay Percent Range >99%
Molecular Formula C15H18N2
CAS 5465-86-1
Chemical Name or Material ML 204
Synonym 4-Methyl-2-(1-piperidinyl)quinoline
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C