Please login to your online account to display your discounted pricing

ML 161, Tocris™

Inhibits PAR1-mediated platelet activation

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material ML 161
Assay Percent Range >98%
Molecular Formula C17H17BrN2O2
Formula Weight 361.23
Synonym 2-Bromo-N-[3-[(1-oxobutyl)amino]phenyl]benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

457050

 
r & d systems
RDS457050
50mg Each for $862.95

457010

 
r & d systems
RDS457010
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 361.23
Quantity 50mg
Synonym 2-Bromo-N-[3-[(1-oxobutyl)amino]phenyl]benzamide
Chemical Name or Material ML 161
Assay Percent Range >98%
Molecular Formula C17H17BrN2O2
CAS 423735-93-7
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C