missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ML 141, Tocris™

Selective inhibitor of Cdc42 Rho family GTPase

$244.81 - $997.23

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C22H21N3O3S
Formula Weight 407.49
Synonym 4-[4, 5-Dihydro-5-(4-methoxyphenyl)-3-phenyl-1H-pyrazol-1-yl]benzenesulfonamide
Chemical Name or Material ML 141
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
426610 R & D Systems
RDS426610
10mg Each for $244.81
426650 R & D Systems
RDS426650
50mg Each for $997.23
Specifications Description & Specifications

Chemical Identifiers

>99%
407.49
ML 141
C22H21N3O3S
4-[4, 5-Dihydro-5-(4-methoxyphenyl)-3-phenyl-1H-pyrazol-1-yl]benzenesulfonamide

Specifications

10mg
>99%
407.49
ML 141
71203-35-5
C22H21N3O3S
4-[4, 5-Dihydro-5-(4-methoxyphenyl)-3-phenyl-1H-pyrazol-1-yl]benzenesulfonamide

Safety and Handling

Recommended Storage : Store at +4°C