missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ML 130, Tocris™

Inhibits NOD1-induced NF-κB activation

$225.77 - $924.23

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C14H13N3O2S
Formula Weight 287.34
Synonym 1-[(4-Methylphenyl)sulfonyl]-1H-benzimidazol-2-amine
Chemical Name or Material ML 130
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
435410 R & D Systems
RDS435410
10mg Each for $225.77
435450 R & D Systems
RDS435450
50mg Each for $924.23
Specifications Description & Specifications

Chemical Identifiers

>99%
287.34
ML 130
C14H13N3O2S
1-[(4-Methylphenyl)sulfonyl]-1H-benzimidazol-2-amine

Specifications

10mg
>99%
287.34
ML 130
799264-47-4
C14H13N3O2S
1-[(4-Methylphenyl)sulfonyl]-1H-benzimidazol-2-amine

Safety and Handling

Recommended Storage : Store at +4°C