Please login to your online account to display your discounted pricing

ML 10302 hydrochloride, Tocris™

Potent and selective 5-HT4 partial agonist

$126.32 - $534.65

Chemical Identifiers

Chemical Name or Material ML 10302 hydrochloride
Assay Percent Range >99%
Molecular Formula C15H21ClN2O3·HCl
Formula Weight 349.25
Synonym 4-Amino-5-chloro-2-methoxybenzoic acid 2-(1-piperidinyl)ethyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

349910

 
r & d systems
RDS349910
10mg Each for $126.32

349950

 
r & d systems
RDS349950
50mg Each for $534.65
Description & Specifications

Specifications

Formula Weight 349.25
Quantity 10mg
Chemical Name or Material ML 10302 hydrochloride
Assay Percent Range >99%
Molecular Formula C15H21ClN2O3·HCl
Recommended Storage Desiccate at Room Temperature
Synonym 4-Amino-5-chloro-2-methoxybenzoic acid 2-(1-piperidinyl)ethyl ester hydrochloride
CAS 186826-17-5
Safety and Handling

Desiccate at Room Temperature