missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MKT 077, Tocris™

Binds mot-2; selectively cytotoxic in cancer cells

$257.85 - $1,073.10

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C21H22ClN3OS2
Formula Weight 432
Synonym 1-Ethyl-2-[[3-ethyl-5-(3-methyl-2(3H)-benzothiazolylidene)-4-oxo-2-thiazolidinylidene]methyl]-pyridinium chloride
Chemical Name or Material MKT 077
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
462110 R & D Systems
RDS462110
10mg Each for $257.85
462150 R & D Systems
RDS462150
50mg Each for $1,073.10
Specifications Description & Specifications

Chemical Identifiers

>98%
432
MKT 077
C21H22ClN3OS2
1-Ethyl-2-[[3-ethyl-5-(3-methyl-2(3H)-benzothiazolylidene)-4-oxo-2-thiazolidinylidene]methyl]-pyridinium chloride

Specifications

147366-41-4
C21H22ClN3OS2
1-Ethyl-2-[[3-ethyl-5-(3-methyl-2(3H)-benzothiazolylidene)-4-oxo-2-thiazolidinylidene]methyl]-pyridinium chloride
MKT 077
>98%
432
10mg

Safety and Handling

Recommended Storage : Store at +4°C