missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ MKNK2 (MNK2) Human Protein DFS Item

MKNK1 and MKNK2 are the Map kinase-interacting kinases.

$574.00 - $8,390.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV5607 View Documents Invitrogen™
LSPV5607
10μg Each for $574.00
 
PR8046A View Documents Invitrogen™
LSPR8046A
100μg Each for $1,976.00
 
PV5608 View Documents Invitrogen™
LSPV5608
1mg Each for $8,390.00
 
Description

Description

The Map kinase-interacting kinases are activated by Map kinases, regulate protein synthesis by phosphroylation eIF4E on Ser209, and may contribute to tumorigenesis.
Specifications

Specifications

Frozen
Human
GST Tag