Invitrogen™ MKNK1 (MNK1) Human Protein DFS Item
MKNK1 and MKNK2 are the Map kinase-interacting kinases.
Manufacturer:  Invitrogen™ LSPV6024
 View more versions of this product
Catalog No. PV6024
Description
Description
The Map kinase-interacting kinases are activated by Map kinases, regulate protein synthesis by phosphroylating eIF4E on Ser209, and may contribute to tumorigenesis.
Specifications
Specifications
NP_945324
Frozen
1mg
RUO - research use only
MKNK1 (MNK1)
Insect
Human
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
66.6kDa
Kinase proteins are shipped on dry ice and should be stored at -80°C.
Full Length
Kinase Assay
MAPKAPK Kinase Family
GST Tag
Documents