Please login to your online account to display your discounted pricing

(+)-MK 801 maleate, Tocris


Non-competitive NMDA antagonist, acts at ion channel site

Quantity 10mg

Manufacturer: r & d systems  092410

 View more versions of this product

Catalog No. 092410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 337.37
Chemical Name or Material (+)-MK 801 maleate
Assay Percent Range >99%
CAS 77086-22-7
Synonym (5S,10R)-(+)-5-Methyl-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5,10-imine maleate
Recommended Storage Store at Room Temperature
Molecular Formula C16H15N·C4H4O4

Specifications

Chemical Name or Material (+)-MK 801 maleate
Assay Percent Range >99%
Molecular Formula C16H15N·C4H4O4
Formula Weight 337.37
Synonym (5S,10R)-(+)-5-Methyl-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5,10-imine maleate
Accessories

Store at Room Temperature