Please login to your online account to display your discounted pricing

MK 212 hydrochloride, Tocris


5-HT2C agonist

Quantity 50mg

Manufacturer: r & d systems  094150

 View more versions of this product

Catalog No. 094150

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material MK 212 hydrochloride
Quantity 50mg
CAS 64022-27-1
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C8H11ClN4·HCl
Formula Weight 235.12
Synonym 6-Chloro-2-(1-piperazinyl)pyrazine hydrochloride

Specifications

Chemical Name or Material MK 212 hydrochloride
Assay Percent Range >99%
Molecular Formula C8H11ClN4·HCl
Formula Weight 235.12
Synonym 6-Chloro-2-(1-piperazinyl)pyrazine hydrochloride
Accessories

Store at Room Temperature