Please login to your online account to display your discounted pricing

MK 0343, Tocris™

Subtype-selective GABAA receptor partial agonist

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material MK 0343
Assay Percent Range >99%
Molecular Formula C19H17F2N7O
Formula Weight 397.38
Synonym 7-Cyclobutyl-3-(2,6-difluorophenyl)-6-[(1-methyl-1H-1,2,4-triazol-5-yl)methoxy]-1,2,4-triazo[4,3-b]pyridazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

441550

 
r & d systems
441550
50mg Each for $1,180.32

441510

 
r & d systems
441510
10mg Each for $284.81
Description & Specifications

Specifications

Formula Weight 397.38
Quantity 50mg
Chemical Name or Material MK 0343
Assay Percent Range >99%
Molecular Formula C19H17F2N7O
CAS 233275-76-8
Synonym 7-Cyclobutyl-3-(2,6-difluorophenyl)-6-[(1-methyl-1H-1,2,4-triazol-5-yl)methoxy]-1,2,4-triazo[4,3-b]pyridazine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C