missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mizoribine, TCI America™

$223.06 - $777.66

Chemical Identifiers

CAS 50924-49-7
Molecular Formula C9H13N3O6
Molecular Weight (g/mol) 259.218
MDL Number MFCD00057221
InChI Key HZQDCMWJEBCWBR-UUOKFMHZSA-N
Synonym 1-(beta-D-Ribofuranosyl)-5-hydroxyimidazole-4-carboxamide
PubChem CID 104762
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-hydroxyimidazole-4-carboxamide
SMILES C1=NC(=C(N1C2C(C(C(O2)CO)O)O)O)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M239950MG View Documents TCI America
M239950MG
50mg Each for $223.06
 
M2399250MG View Documents TCI America
M2399250MG
250mg Each for $777.66
 
Specifications

Chemical Identifiers

50924-49-7
259.218
HZQDCMWJEBCWBR-UUOKFMHZSA-N
104762
C1=NC(=C(N1C2C(C(C(O2)CO)O)O)O)C(=O)N
C9H13N3O6
MFCD00057221
1-(beta-D-Ribofuranosyl)-5-hydroxyimidazole-4-carboxamide
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-hydroxyimidazole-4-carboxamide

Specifications

White-Yellow
50mg
C9H13N3O6
1-(beta-D-Ribofuranosyl)-5-hydroxyimidazole-4-carboxamide
C1=NC(=C(N1C2C(C(C(O2)CO)O)O)O)C(=O)N
259.218
259.22
≥98.0% (HPLC)
198°C
50924-49-7
MFCD00057221
HZQDCMWJEBCWBR-UUOKFMHZSA-N
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-hydroxyimidazole-4-carboxamide
104762
Crystal-Powder at 20°C
Mizoribine
Safety and Handling

Safety and Handling

RTECSNumber : NI3980000

TSCA : No

Recommended Storage : Refrigerator