Please login to your online account to display your discounted pricing

Mixed Ind. Soln, Methyl Red - Methylene Blue, APHA/EPA for Nitrogen, Ricca Chemical

$46.09 - $87.18

Chemical Identifiers

Chemical Name or Material Mixed Indicator Solution
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5164-16

 
ricca chemical company
RI516416
Plastic 500mL Each for $87.18

5164-4

 
ricca chemical company
RI51644
Plastic 120mL Each for $46.09
Description & Specifications

Specifications

Quantity 500mL
Methods APHA/EPA for Nitrogen
Concentration or Composition (by Analyte or Components) Methyl Red - Methylene Blue
Packaging Plastic
Chemical Name or Material Mixed Indicator Solution