Please login to your online account to display your discounted pricing

Mixed Calibration 4, Ricca Chemical


Quantity 100mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIRMXCL4100

 View more versions of this product

Catalog No. RMXCL4-100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 100mL
Color Clear colorless
Odor Odorless
Chemical Name or Material Mixed Calibration 4
CAS 7697-37-2 (Nitric Acid)
Shelf Life 12 months
CAS 7732-18-5 (Water, Deionized)
Packaging Poly natural

Specifications

Chemical Name or Material Mixed Calibration 4
CAS 7697-37-2 (Nitric Acid)
CAS 7732-18-5 (Water, Deionized)
Accessories

12 months