Please login to your online account to display your discounted pricing

Mitosis and Meiosis Lab Investigations


An exciting and interactive way to learn about mitosis and meiosis.

Manufacturer: fisher science education  AZ-20-1283

 View more versions of this product

Catalog No. S32617

  • / Each
 

Description & Specifications

Specifications

Product Type Meiosis Simulation Lab Investigation Kit
Kit Size 40 students
For Use With (Application) Students learn about the formation of sex cells
Class Size 40 students