Please login to your online account to display your discounted pricing

Mitosis and Meiosis Lab Investigations


An exciting and interactive way to learn about mitosis and meiosis.

Manufacturer: fisher science education  AZ-20-2183

 View more versions of this product

Catalog No. S65122

  • / Each
 

Description & Specifications

Specifications

Kit Size 40 students
For Use With (Application) Helps to visualize the process of mitosis
Product Type Mitosis Simulation Lab Investigation Kit
Class Size 40 students