Please login to your online account to display your discounted pricing

Alfa Aesar™ Mitomycin C, 5mg/mL in ethanol, sterile-filtered

Chemical Identifiers

Chemical Name or Material Mitomycin C
CAS 50-07-7
Name Note 5mg/mL in ethanol, sterile-filtered
MDL Number MFCD00078109
UN Number UN1987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67460XF

  SDS
alfa aesar™
J67460XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Mitomycin C
Health Hazard 2 GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.
Physical Form Liquid
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
CAS 50-07-7
RTECS Number CN0700000
TSCA TSCA
Storage Note 1 -30° to -10°C
Quantity 1mL
Name Note 5mg/mL in ethanol, sterile-filtered
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
UN Number UN1987
MDL Number MFCD00078109
Safety and Handling

Danger

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.

-30° to -10°C