Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mitochondria Ab-2, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mitochondria Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1372P1ABX

 View more versions of this product

Catalog No. MS1372P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Isotype IgG1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Semi-purified mitochondrial preparation
Clone MTC02
Cross Reactivity Human
Regulatory Status RUO
Quantity 100µL
Antigen Mitochondria Ab-2
Format Concentrated
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Cell Cycle
Formulation Ab Without BSA/Azide (Frozen)
Type Primary
Concentration 1mg/mL

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mitochondria Ab-2, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MTC02

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified mitochondrial preparation

Molecular Weight: 60kDa

Positive Control: HeLa cells, Liver

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO