missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mitochondria Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mitochondria Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1372PABX

 View more versions of this product

Catalog No. MS1372PABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mitochondria Ab-2, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MTC02

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified mitochondrial preparation

Molecular Weight: 60kDa

Positive Control: HeLa cells, Liver

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Mitochondria Ab-2
Monoclonal
1mg/mL
Human
Ab Without BSA/Azide (Frozen)
Semi-purified mitochondrial preparation
200μL
Cell Cycle
Monoclonal
Immunofluorescence,Immunohistochemistry (Paraffin),Western Blotting
MTC02
Unlabeled
Concentrated
Murine
IgG1
RUO
Primary
Human