Please login to your online account to display your discounted pricing

MitMAB, Tocris™

Dynamin inhibitor

$75.29 - $317.73

Chemical Identifiers

Chemical Name or Material MitMAB
Assay Percent Range >98%
Molecular Formula C17H38BrN
Formula Weight 336.39
Synonym Tetradecyltrimethylammonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

422410

 
r & d systems
RDS422410
10mg Each for $75.29

422450

 
r & d systems
RDS422450
50mg Each for $317.73
Description & Specifications

Specifications

Chemical Name or Material MitMAB
Quantity 10mg
CAS 1119-97-7
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >98%
Molecular Formula C17H38BrN
Formula Weight 336.39
Synonym Tetradecyltrimethylammonium bromide
Safety and Handling

Desiccate at Room Temperature