Please login to your online account to display your discounted pricing

Alfa Aesar™ Misoprostol, free acid


Quantity 5mg

Manufacturer: alfa aesar™  J67475LB0

 View more versions of this product

Catalog No. AAJ67475LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 368.51
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Misoprostol
Name Note Free acid
Physical Form Liquid
CAS 112137-89-0
Health Hazard 2 GHS H Statement
H301-H360-H315-H319-H335
Toxic if swallowed.
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
MDL Number MFCD00869984
UN Number UN2810
Health Hazard 1 Danger
Color Yellow
Quantity 5mg
Storage Note 1 -30° to -10°C

Specifications

Chemical Name or Material Misoprostol
CAS 112137-89-0
Name Note Free acid
Formula Weight 368.51
MDL Number MFCD00869984
UN Number UN2810
Accessories

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H360-H315-H319-H335
Toxic if swallowed.
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C

SDS & Safety Documents