Please login to your online account to display your discounted pricing

Alfa Aesar™ Misch Metal, 99.8% min. rare earth content, trapezoidal ingot, dryb

Chemical Identifiers

Chemical Name or Material Misch Metal
CAS 62379-61-7
Name Note Dry
MDL Number MFCD00144799
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA66144P2

  SDS
alfa aesar™
66144P2
1 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 lb.
Assay Percent Notes rare earth content
Concentration Ce rich (varies) 55 to 70%
Chemical Name or Material Misch Metal
TSCA TSCA
Assay ≥99.8%
Odor Odorless
Physical Form Trapezoid Ingot
DOT Information Transport Hazard Class: 4.1; Packaging Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Melting Point ≈845°C
Health Hazard 1 Danger
MDL Number MFCD00144799
Sensitivity Air & Moisture Sensitive
CAS 62379-61-7
Health Hazard 2 GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
UN Number UN3178
Health Hazard 3 GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Silver
Weight ≈500g/ingot
Name Note Dry
Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.