missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Misch Metal, 99.0% min. rare earth content

Manufacturer:  Alfa Aesar™ 45589A3

 View more versions of this product

Catalog No. AA45589A3



Specifications

Specifications

Typically Ce:La 75:25 wt%
Air & Moisture Sensitive
62379-61-7
MFCD00144799
misch metal, ce.la.nd.pr, cerium lanthanum neodymium praseodymium
≥99.0%
Misch Metal
2kg
rare earth content
UN3178
Pieces

Chemical Identifiers

62379-61-7
MFCD00144799
misch metal, ce.la.nd.pr, cerium lanthanum neodymium praseodymium
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents