Please login to your online account to display your discounted pricing

Mirtazapine, Tocris

Potent 5-HT2 antagonist; Also 5-HT3, H1 and α2-antagonist; Antidepressant

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material Mirtazapine
Assay Percent Range >99%
Molecular Formula C17H19N3
Formula Weight 265.36
Synonym 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

201810

 
r & d systems
201810
10mg Each for $142.08

201850

 
r & d systems
201850
50mg Each for $598.33
Description & Specifications

Specifications

Formula Weight 265.36
Quantity 10mg
Chemical Name or Material Mirtazapine
Assay Percent Range >99%
Molecular Formula C17H19N3
CAS 85650-52-8
Synonym 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature