Mirror Assortment
Manufacturer: Fisher Science Education OPK10
Description
A quality assortment of aluminum or dielectric mirrors!
A quality assortment of different types of new front-surface aluminum or dielectric mirrors of various sizes. Please note that some mirrors have a protective layer over the first surface. Peel off before use.
Order Info
Sizes and focal lengths vary in each package; 10 pieces per assortment.
Specifications
Aluminum | |
Mirror assortment | |
Each |