missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mirin, Tocris

MRN-ATM pathway inhibitor

$187.24 - $789.24

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C10H8N2O2S
Formula Weight 220.25
Synonym Z-5-(4-Hydroxybenzylidene)-2-imino-1, 3-thiazolidin-4-one
Chemical Name or Material Mirin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
319010 R & D Systems
RDS319010
10mg Each for $187.24
319050 R & D Systems
RDS319050
50mg Each for $789.24
Specifications Description & Specifications

Chemical Identifiers

>99%
220.25
Mirin
C10H8N2O2S
Z-5-(4-Hydroxybenzylidene)-2-imino-1, 3-thiazolidin-4-one

Specifications

1198097-97-0
C10H8N2O2S
Z-5-(4-Hydroxybenzylidene)-2-imino-1, 3-thiazolidin-4-one
Mirin
>99%
220.25
10mg

Safety and Handling

Recommended Storage : Store at +4°C