Please login to your online account to display your discounted pricing

MIRA-1, Tocris

Restores mutant p53 activity; proapoptotic

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material MIRA-1
Assay Percent Range >98%
Molecular Formula C8H9NO4
Formula Weight 183.16
Synonym 1-[(1-Oxopropoxy)methyl]-1H-pyrrole-2, 5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

336210

 
r & d systems
336210
10mg Each for $139.08

336250

 
r & d systems
336250
50mg Each for $585.69
Description & Specifications

Specifications

Formula Weight 183.16
Quantity 10mg
Chemical Name or Material MIRA-1
Assay Percent Range >98%
Synonym 1-[(1-Oxopropoxy)methyl]-1H-pyrrole-2, 5-dione
Molecular Formula C8H9NO4
CAS 72835-26-8
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C