missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minuteman™ X829 Series HEPA Vacuums
Safe, easy-to-use vacuum for difficult dry and wet cleaning jobs
$2508.99 - $2508.99
Specifications
For Use With (Application) | Hazardous Dusts |
---|---|
Material | Polyethylene |
Filter Type | HEPA |
Efficiency | 99.99% |
Catalog Number | Mfr. No. | Capacity (Metric) | Capacity (Metric) Tank | Product Type | Voltage | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Capacity (Metric) Tank | Product Type | Voltage | Price | Quantity | ||
18-999-985
|
Minuteman™
C8291506 |
Wet: 49 L and Dry: 0.23 cu. m. | 57 L | X829 series HEPA vacuum Wet or Dry | 115/220 V |
Each for $2,508.99
|
|
||
Description
Patented safety interlock system prevents operation if filter is incorrectly aligned.
- Removes hazardous dusts such as lead, arsenic, and asbestos, and nonhazardous wastes such as flour, wood, and foundry dusts
- Durable tank has a quick-release, self-sealing intake that ensures a positive vacuum seal
- ULPA (Ultra Low Penetration Air) filter is 99.999% efficient at 0.12μm; resists punctures, tears, leaks, abrasions, and moisture damage
- Patented safety interlock system ensures the ULPA filter is properly in place
- One-piece, polyethylene Uni-Adapter™ attaches easily to form a leakproof seal with the vacuum tank, enabling safe wet direct load recovery
Order Info
15 gal. (57L) tank has a wet capacity of 13 gal. (49L) and a dry capacity of 0.82 cu. ft. (0.023m3). Decibel rating is 72dB.
Specifications
Hazardous Dusts | |
Polyethylene | |
HEPA | |
99.99% |