Please login to your online account to display your discounted pricing
Minuteman™ Allergy Vacuum
Four-stage filtration system contains common allergens
$859.85 - $859.85
Specifications
Filter Type HEPA
View More Specs
Includes:  Caddy and six-piece tool kit containing a combination floor tool, crevice tool, dust brush, upholstery tool, two-piece wand and 1.25 in. dia. (3.2cm) hose.
Products
Catalog Number Mfr. No. Product Type Price Quantity    
19-063-070
minuteman international
C8290407
Allergy vacuum Each for $859.85
Description & Specifications
Specifications
Filter Type HEPA
  • Captures common allergens, such as pollen, mold, animal dander, and dust mites that collect in carpeting, draperies, bedding and furniture
  • Traps harmful allergens with progressively finer stages of filtration
  • Four-stage filtration system consists of a disposable paper bag, a washable embossed polyester filter bag, a disposable impact filter and a certified HEPA (High-Efficiency Particulate Air) filter
  • HEPA Filter is 99% efficient at 0.3 microns; pnk impaction filter is 95% efficient at 0.3 microns; embossed polyester filter bag is 99% efficient at 1.5 microns; and disposable paper bag is 99% efficient at 5.0 microns
  • Patented safety interlock system assures user safety and prevents allergens for being redistributed back into the air; vacuum will not operate if HEPA filter is removed
  • 4 gal. (15L) polyethylene tank is noncorrosive and dent-proof with four 2 in. dia. (5cm) casters, patented self sealing intake, and 40 foot cord
Power: 1.25hp (.93 kw). Dry capacity: 0.21 cu. ft. (0.006m3). Airflow: 95cfm (2.69m3). Static lift: 85 in. (21.6cm). Sound rating: 68 dBA. Weight: 16 lb. (7kg). Replacement HEPA filters sold separately.