Minuteman™ Allergy Vacuum
Four-stage filtration system contains common allergens
Manufacturer: Minuteman™ C8290407
Description
- Captures common allergens, such as pollen, mold, animal dander, and dust mites that collect in carpeting, draperies, bedding and furniture
- Traps harmful allergens with progressively finer stages of filtration
- Four-stage filtration system consists of a disposable paper bag, a washable embossed polyester filter bag, a disposable impact filter and a certified HEPA (High-Efficiency Particulate Air) filter
- HEPA Filter is 99% efficient at 0.3 microns; pnk impaction filter is 95% efficient at 0.3 microns; embossed polyester filter bag is 99% efficient at 1.5 microns; and disposable paper bag is 99% efficient at 5.0 microns
- Patented safety interlock system assures user safety and prevents allergens for being redistributed back into the air; vacuum will not operate if HEPA filter is removed
- 4 gal. (15L) polyethylene tank is noncorrosive and dent-proof with four 2 in. dia. (5cm) casters, patented self sealing intake, and 40 foot cord
Specifications
Disposable collector bag, pre filter and HEPA filter, 1.25 in. x 8 ft. hose, telescopic operator handle, crevice, dust brush, combo floor tools and collector bags | |
0.34 cu. ft. | |
HEPA | |
Polyethylene | |
115V | |
0.93w | |
35kg | |
15L |
0.01m3/min. | |
Allergy vacuum | |
1.25hp | |
68dB | |
85 in. | |
77 lb. | |
512 oz. | |
0.34 cu. ft., 0.01m3/min. |