Learn More
Minocycline hydrochloride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J66429ME
Description
A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections
Specifications
Minocycline Hydrochloride | |
890 to 950μg/mg | |
13614-98-7 | |
C23H28ClN3O7 | |
14,6202 | |
KDLQIOPKJDNQIM-WUURTAMISA-N | |
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride | |
54685925 | |
Yellow | |
>110°C (230°F) |
Minocycline hydrochloride | |
Powder | |
205°C to 210°C | |
MFCD00083669 | |
Freely soluble in water | |
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C | |
493.94 | |
493.94 | |
500 mg | |
813°C |
Chemical Identifiers
13614-98-7 | |
493.94 | |
KDLQIOPKJDNQIM-WUURTAMISA-N | |
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride |
C23H28ClN3O7 | |
MFCD00083669 | |
54685925 | |
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 237-099-7
RTECSNumber : QI7630500
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only