missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Minocycline hydrochloride, 50mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Manufacturer:  Alfa Aesar J674698EQ

 View more versions of this product

Catalog No. AAJ674698EQ



Specifications

Specifications

Minocycline hydrochloride
13614-98-7
MFCD00083669
CN(C)C1C2CC3CC4=C(C=CC(=C4C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O)N(C)C.Cl
493.941
Liquid
5 x 1mL
sterile-filtered
C23H28ClN3O7
WTJXVDPDEQKTCV-VQAITOIOSA-N
(4S,4aS,5aR,12aR)-4,7-bis(dimethylamino)-1,10,11,12a-tetrahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
54685925
50mg/mL in distilled water

Chemical Identifiers

13614-98-7
493.941
WTJXVDPDEQKTCV-VQAITOIOSA-N
(4S,4aS,5aR,12aR)-4,7-bis(dimethylamino)-1,10,11,12a-tetrahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
C23H28ClN3O7
MFCD00083669
54685925
CN(C)C1C2CC3CC4=C(C=CC(=C4C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O)N(C)C.Cl
Safety and Handling

Safety and Handling

RTECSNumber : QI7630500

storageNote1 : −30° to −10°C

Documents