Invitrogen™ MINK1 Recombinant Human Protein DFS Item
MINK1 is a serine/threonine kinase belonging to the germinal center kinase (GCK) family.
Manufacturer:  Invitrogen™ LSPV4022
 View more versions of this product
Catalog No. PV4022
Specifications
Specifications
NP_056531
Frozen
63.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
STE20 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MINK1, MINK
Baculovirus
Human
GST Tag
Documents