Please login to your online account to display your discounted pricing

Mineral Oil USP White, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Mineral Oil
Name Note Heavy
CAS 8012-95-1
Purity Grade USP
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK63574

 
avantor performance
635710
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color White
Quantity 4L
Chemical Name or Material Mineral Oil
Name Note Heavy
Purity Grade USP
CAS 8012-95-1
Packaging Glass