Learn More
Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.
CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Supplier: Thermo Scientific Chemicals 124020025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For IR spectroscopySpecifications
| Mineral oil | |
| Colorless | |
| 185°C | |
| Glass Bottle | |
| 1.4620 to 1.4730 | |
| 2.5 L | |
| 14, 7186 | |
| Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether,, carbon dislulfide, petroleum ether, oils, miscible with wax and fats, insoluble in alcohol | |
| C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] | |
| 452.363 | |
| 14-19 mm2/s (40°C) | |
| Oily Liquid |
| 8042-47-5 | |
| 0.8510g/mL | |
| Authentic | |
| C16H10N2Na2O7S2 | |
| MFCD00131611 | |
| 0.851 | |
| acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g | |
| AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
| disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate | |
| 9566064 | |
| Spectroscopy |
Chemical Identifiers
| 8042-47-5 | |
| 452.363 | |
| AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
| 9566064 | |
| C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
| C16H10N2Na2O7S2 | |
| MFCD00131611 | |
| acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g | |
| disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 232-455-8
RTECSNumber : PY8047000
TSCA : TSCA
RUO – Research Use Only