missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect., ACROS Organics™

Manufacturer:  Fisher Scientific Acros 124020060

 View more versions of this product

Catalog No. AC124020060



Description

Description

For IR spectroscopy
Specifications

Specifications

Mineraloil
185°C
Glass Bottle
0.851
Colorless
8042-47-5
MFCD00131611
acid orange 10, c.i. acid orange 10, c.i. food orange 4, c.i. orange g, colacid orange g, dolkwal orange g, hexacol orange g, light orange g, orange g, wool orange 2g
AEOVEGJBKQQFOP-DDVLFWKVSA-L
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
9566064
Spectroscopy
0.8510g/mL
Authentic
1.4620 to 1.4730
14-19 mm2/s (40°C)
6mL
C16H10N2Na2O7S2
14, 7186
Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether,, carbon dislulfide, petroleum ether, oils, miscible with wax and fats, insoluble in alcohol
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
452.363
Oily Liquid

Chemical Identifiers

8042-47-5
452.363
AEOVEGJBKQQFOP-DDVLFWKVSA-L
9566064
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C16H10N2Na2O7S2
MFCD00131611
acid orange 10, c.i. acid orange 10, c.i. food orange 4, c.i. orange g, colacid orange g, dolkwal orange g, hexacol orange g, light orange g, orange g, wool orange 2g
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 232-455-8

RTECSNumber : PY8047000

TSCA : TSCA

SDS Documents