missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mineral oil, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 415080010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Mineral oil | |
Colorless | |
220°C | |
Glass bottle | |
1.4760 to 1.4830 | |
0.877 | |
Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether, cs2,, petroleum ether, oils, miscible with wax and fats, insoluble in ethanol | |
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] | |
452.363 | |
65-75 mm2/s (40°C) | |
Oily Liquid |
8042-47-5 | |
0.8770g/mL | |
Authentic | |
C16H10N2Na2O7S2 | |
1 L | |
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g | |
AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate | |
9566064 | |
Pure |
Chemical Identifiers
8042-47-5 | |
452.363 | |
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g | |
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate |
C16H10N2Na2O7S2 | |
AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
9566064 | |
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
Safety and Handling
EINECSNumber : 232-455-8
RUO – Research Use Only