missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mineral Oil, Light (NF/FCC), Fisher Chemical

Saybolt viscosity: 158 maximum

Manufacturer:  Thermo Scientific O1214

 View more versions of this product

Catalog No. O121-4




Specifications

Specifications

Pass Test (FCC)
Mineral Oil, Light
160°C
Glass Bottle
0.818 to 0.880 (FCC/NF)
3 CS min. (FCC), 3.0 to 34.4 mm2.S-1 (NF)
White
8042-47-5
MFCD00131611
AEOVEGJBKQQFOP-DDVLFWKVSA-L
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
9566064
FCC/NF
Pass Test (NF)
0.83g/mL
Pass Test (FCC/NF)
Meets Requirements (NF)
<0.1 mmHg
260°C
4L
C16H10N2Na2O7S2
acid orange 10
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
452.363
Liquid

Chemical Identifiers

8042-47-5
452.363
AEOVEGJBKQQFOP-DDVLFWKVSA-L
9566064
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C16H10N2Na2O7S2
MFCD00131611
acid orange 10
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:0
Flammability:1
Instability:0

SDS Documents
Accessories