Please login to your online account to display your discounted pricing

MIM1, Tocris™

Mcl-1 inhibitor; proapoptotic

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material MIM1
Assay Percent Range >98%
Molecular Formula C17H21N3O3S
Formula Weight 347.43
Synonym 4-(E)-[[(2Z)-2-cyclohexylimino)-4-methyl-1,3-thiazol-3(2H)-yl]imino]methyl]benzene-1,2,3-triol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

476210

 
r & d systems
RDS476210
10mg Each for $221.01

476250

 
r & d systems
RDS476250
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 347.43
Quantity 10mg
Chemical Name or Material MIM1
Molecular Formula C17H21N3O3S
Assay Percent Range >98%
Synonym 4-(E)-[[(2Z)-2-cyclohexylimino)-4-methyl-1,3-thiazol-3(2H)-yl]imino]methyl]benzene-1,2,3-triol
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C